BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Ciprofloxacin31
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
$27.63 - $128.21
Specifications
Antimicrobial Agents | Ciprofloxacin31 |
---|---|
Concentration | 5μg |
Packaging Type | Disc |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
B31657
|
BD
231657 |
1 Pack |
Each for $27.63
|
|
|||||
B31658
|
BD
231658 |
10 Pack |
Case of 10 Each for $128.21
|
|
|||||
Description
- 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
- Are clearly marked on both sides with letters and numbers designating agent and drug content
Specifications
Ciprofloxacin31 | |
5μg | |
Disc |
For in vitro diagnostic use