missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BD Difco™ Dehydrated Culture Media: Fraser Broth Base
Use with Fraser Broth Supplement Selective enrichment and detection of Listeria
Supplier: BD 211767
Description
- Formulation: Pancreatic digest of Casein 5g/L, Proteose Peptone No.3 5g/L, Beef extract 5g/L, Yeast extract 5g/L, Sodium Chloride 20g/L, Disodium Phosphate 9.6g/L, Monopotassium Phosphate 1.35g/L, Esculin 1g/L, Nalidixic Acid 0.02g/L, Acriflavine HCl 24mg/L, Lithium Chloride 3g/L
Certifications
CCAM, COMPF, ISO, USDA
Specifications
CCAM, COMPF, ISO, USDA | |
Dehydrated | |
Microbiology Media | |
Used with Fraser broth supplement selective enrichment and detection of Listeria | |
500 g |
2°C to 8°C | |
Fraser Broth Base | |
Pancreatic digest of Casein 5g/L, Proteose Peptone No.3 5g/L, Beef extract 5g/L, Yeast extract 5g/L, Sodium Chloride 20g/L, Disodium Phosphate 9.6g/L, Monopotassium Phosphate 1.35g/L, Esculin 1g/L, Nalidixic Acid 0.02g/L, Acriflavine HCl 24mg/L, Lithium Chloride 3g/L | |
Bottle |