missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BD Difco™ Dehydrated Culture Media: Fraser Broth Base
Use with Fraser Broth Supplement Selective enrichment and detection of Listeria
$292.16 - $1215.58
Specifications
Certifications/Compliance | CCAM, COMPF, ISO, USDA |
---|---|
Content And Storage | 2°C to 8°C |
Form | Dehydrated |
Media Type | Fraser Broth Base |
Product Type | Microbiology Media |
Catalog Number | Mfr. No. | Quantity | Packaging Type | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging Type | Price | Quantity | ||||
DF0219-07-0
|
BD
211766 |
2000 g | Pail |
Each for $1,215.58
|
|
||||
DF0219-17-8
|
BD
211767 |
500 g | Bottle |
Each for $292.16
|
|
||||
Description
- Formulation: Pancreatic digest of Casein 5g/L, Proteose Peptone No.3 5g/L, Beef extract 5g/L, Yeast extract 5g/L, Sodium Chloride 20g/L, Disodium Phosphate 9.6g/L, Monopotassium Phosphate 1.35g/L, Esculin 1g/L, Nalidixic Acid 0.02g/L, Acriflavine HCl 24mg/L, Lithium Chloride 3g/L
Certifications
CCAM, COMPF, ISO, USDA
Specifications
CCAM, COMPF, ISO, USDA | |
Dehydrated | |
Microbiology Media | |
Used with Fraser broth supplement selective enrichment and detection of Listeria |
2°C to 8°C | |
Fraser Broth Base | |
Pancreatic digest of Casein 5g/L, Proteose Peptone No.3 5g/L, Beef extract 5g/L, Yeast extract 5g/L, Sodium Chloride 20g/L, Disodium Phosphate 9.6g/L, Monopotassium Phosphate 1.35g/L, Esculin 1g/L, Nalidixic Acid 0.02g/L, Acriflavine HCl 24mg/L, Lithium Chloride 3g/L |