BD General Use and PrecisionGlide Hypodermic Needles
Fit any Luer-Lok™, slip tip, or eccentric tip syringe
$13.93 - $42.50
Specifications
Product Type | Conventional Hypodermic Needle |
---|---|
Material | Stainless Steel |
Sterility | Sterile |
Cannula Material | Siliconized |
Connectors | Luer-Lok |
Catalog Number | Mfr. No. | Diameter (English) Outside | Gauge | Length (English) | Thickness (Wall) | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (English) Outside | Gauge | Length (English) | Thickness (Wall) | Price | Quantity | ||
14-826-18A
|
BD
305197 |
0.064 in. | 16 G | 1 in. | Regular |
Pack of 100 Each for $39.48
Case of 10 Pack for $394.80
|
|
||
14-826-18B
|
BD
305198 |
0.064 in. | 16 G | 1.5 in. | Regular |
Pack of 100 Each for $42.50
Case of 10 Pack for $425.00
|
|
||
14-826-49
|
BD
305127 |
0.02 in. | 25 G | 1.5 in. | Regular |
Pack of 100 Each for $14.89
Case of 10 Pack for $148.90
|
|
||
14-826-5G
|
BD
305195 |
0.048 in. | 18 G | 1 in. | Regular |
Pack of 100 Each for $13.93
Case of 10 Pack for $139.30
|
|
||
14-826-6C
|
BD
305194 |
0.025 in. | 23 G | 1.5 in. | Thin |
Pack of 100 Each for $14.89
Case of 10 Pack for $148.90
|
|
||
14-826C
|
BD
305165 |
0.031 in. | 21 G | 1 in. | Regular |
Pack of 100 Each for $14.89
Case of 10 Pack for $148.90
|
|
||
14-826-48
|
BD
305109 |
0.016 in. | 27 G | 0.5 in. | Regular |
Pack of 100 Each for $19.34
Case of 10 Pack for $193.40
|
|
||
14-826-5B
|
BD
305167 |
0.031 in. | 21 G | 1.5 in. | Regular |
Pack of 100 Each for $14.89
Case of 10 Pack for $148.90
|
|
||
14-826-5D
|
BD
305196 |
0.048 in. | 18 G | 1.5 in. | Regular |
Pack of 100 Each for $13.93
Case of 10 Pack for $139.30
|
|
||
14-826A
|
BD
305145 |
0.025 in. | 23 G | 1 in. | Regular |
Pack of 100 Each for $14.89
Case of 10 Pack for $148.90
|
|
||
14-826AA
|
BD
305122 |
0.02 in. | 25 G | 0.62 in. | Regular |
Pack of 100 Each for $14.89
Case of 10 Pack for $148.90
|
|
||
14-826B
|
BD
305155 |
0.027 in. | 22 G | 1 in. | Regular |
Pack of 100 Each for $14.89
Case of 10 Pack for $148.90
|
|
||
Description
Compatible with any slip tip, Luer-Lok, or eccentric tip syringe.
- Sheath protects point and cannula
- Cannula are siliconized to reduce resistance and maintain speed and smoothness throughout injection
- All have regular bevel, except 14-826-10, which has intradermal bevel
- 14-826-52, -6A, -6B, and -6C needles have thin-wall design
- 14-826-6A is for subcutaneous use and has the same flowrate as larger 25-gauge needle
- 14-826-6B and -6C are for intramuscular use and have the same flowrate as larger 22-gauge needle
- 14-826-52 has the same flowrate as larger 18-gauge needle
- Needle hub fits any Luer-Lok or Luer slip syringe tip, reusable or disposable
- Individual Easy Peel™ blister pack packages boxed in strips of five needles; package carries lot control number Hubs color coded by gauge size: Gray (27-gauge), Tan (26), Blue (25), Turquoise (23), Black (22), Green (21), Yellow (20), Brown (19), Pink (18), and Purple (16)
Order Info
Package carries lot control number to indicate complete sterility.
Specifications
Conventional Hypodermic Needle | |
Sterile | |
Luer-Lok | |
Single-use needles minimize cross-infection and provide a factory-sharp point for each injection | |
Polypropylene | |
Yes |
Stainless Steel | |
Siliconized | |
Single-use | |
Any Luer-Lok, Slip Tip, or Eccentric Tip Syringe | |
Protective Plastic Sheath | |
Bevel Point |