BD Macro-Vue™ RPR Cards and Components
For large-scale testing in public health and clinical laboratories
Supplier: BD 276709
Description
RPR Card antigen suspension is a carbon particle cardiolipin antigen1 which detects reagin, an antibody-like substance present in serum or plasma from syphilitic persons, and occasionally in serum or plasma of persons with other acute or chronic conditions. The reagin binds to the test antigen, which consists of cardiolipin-lecithin-coated cholesterol particles, causing macroscopic flocculation.
- CLIA Complexity: Moderate
- Nontreponemal test
- For the serological detection of syphilis as described in A Manual of Tests for Syphilis†
Specifications
Reagin (Syphilis) | |
For large-scale testing in public health and clinical laboratories | |
10 Pack |
RPR Control Cards | |
BD Macro-Vue RPR control cards, 18-mm circle |