BD Macro-Vue™ RPR Cards and Components
For large-scale testing in public health and clinical laboratories
$133.32 - $934.05
Specifications
For Use With (Application) | For large-scale testing in public health and clinical laboratories |
---|
Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity | ||||
B4970509
|
BD
270509 |
10 Pack | RPR Antigen Dispensing Bottles |
Pack of 10 Each for $242.21
|
|
||||
B4971849
|
BD
271849 |
300 Case | BD RPR Cards |
Case of 300 Each for $934.05
|
|
||||
B4972001
|
BD
272001 |
100 Pack | RPR BD Cards |
Pack of 100 Each for $485.55
|
|
||||
22-130989
|
BD
273509 |
10 Pack | Dispensing Needles (Square Cut), 20 gauge |
Pack of 10 Each for $133.32
|
|
||||
B4976909
|
BD
276909 |
1.5 mL | Liquid RPR Controls |
Each for $191.92
|
|
||||
22-727891
|
BD
276709 |
10 Pack | RPR Control Cards |
Pack of 10 Each for $278.11
|
|
||||
Description
RPR Card antigen suspension is a carbon particle cardiolipin antigen1 which detects reagin, an antibody-like substance present in serum or plasma from syphilitic persons, and occasionally in serum or plasma of persons with other acute or chronic conditions. The reagin binds to the test antigen, which consists of cardiolipin-lecithin-coated cholesterol particles, causing macroscopic flocculation.
- CLIA Complexity: Moderate
- Nontreponemal test
- For the serological detection of syphilis as described in A Manual of Tests for Syphilis†
Specifications
For large-scale testing in public health and clinical laboratories |