BD Microtainer™ Capillary Blood Collector and BD Microgard™ Closure
Textured inner surface and large tube diameter to aid sample mixing
Supplier:  BD 365965
 View more versions of this product
Catalog No. 13-680-62
Explore available promotions
Description
Description
  • Designed to minimize sample contact and improve mixing
  • “Twist assist” feature allows controlled closure removal; snap-lock fit assures complete seal
  • Color-coded closures for accurate tube selection
  • 8mm dia. for easy probe and pipet access; integral collector eliminates need for assembly
  • Marked with fill lines, lot numbers and expiration date.
  • Holds 250 to 500μL of blood
  • Extender (optional) facilitates labeling and handling
Specifications
Specifications
Plasma Tube
Microgard
8 mm
Lithium Heparin
200 to 400 μL
Green