BD Microtainer™ Capillary Blood Collector and BD Microgard™ Closure
Textured inner surface and large tube diameter to aid sample mixing
$58.48 - $586.57
Catalog Number | Mfr. No. | Closure Color | Product Type | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Closure Color | Product Type | Price | Quantity | ||||
02-669-34
![]() |
BD
365976 |
Gray | Tube Extender |
Pack of 50 Each for $58.48
Case of 4 Pack for $233.92
|
![]() |
||||
02-675-184
|
BD
365963 |
Red | Serum Tube |
Case of 200 Each for $586.57
|
|
||||
02-675-185
|
BD
365967 |
Gold | Serum Separator Tube |
Pack of 50 Each for $171.82
Case of 4 Pack for $687.28
|
|
||||
02-675-186
|
BD
365978 |
Gold | Serum Separator Tube |
Pack of 50 Each for $190.64
Case of 4 Pack for $762.56
|
|
||||
02-675-187
|
BD
365985 |
Mint Green | Lithium Heparin with Plasma Separator Tube |
Pack of 50 Each for $146.64
Case of 4 Pack for $586.56
|
|
||||
13-680-62
|
BD
365965 |
Green | Plasma Tube |
Pack of 50 Each for $124.93
Case of 4 Pack for $499.72
|
|
||||
02-669-33
![]() |
BD
365974 |
Lavender | Dipotassium EDTA Tube |
Pack of 50 Each for $325.63
Case of 4 Pack for $1,302.52
|
![]() |
||||
Description
- Designed to minimize sample contact and improve mixing
- “Twist assist” feature allows controlled closure removal; snap-lock fit assures complete seal
- Color-coded closures for accurate tube selection
- 8mm dia. for easy probe and pipet access; integral collector eliminates need for assembly
- Marked with fill lines, lot numbers and expiration date
- Holds 250 to 500μL of blood
- Extender (optional) facilitates labeling and handling
Specifications
Microgard | |
8 mm |