BD Sterile Flocked Swabs
Sterile swabs with soft nylon fiber tip
Supplier: BD 220250
Description
- Swab has no internal mattress core to trap liquid so sample stays closer to surface, allowing easier elution
- Flocked nylon helps dislodge, collect, and transfer solid and semi-solid samples
Order Info
Single wrapped
Specifications
Regular flocked | |
Sterile | |
Sample collection and optimal diagnostics |
Nylon Fiber | |
Individually-wrapped | |
Flocked Swab |