BD Sterile Flocked Swabs
Sterile swabs with soft nylon fiber tip
Supplier:  BD 220252
 View more versions of this product
Catalog No. 22-349-822
Explore available promotions
Description
Description
  • Swab has no internal mattress core to trap liquid so sample stays closer to surface, allowing easier elution
  • Flocked nylon helps dislodge, collect, and transfer solid and semi-solid samples
Order Info
Single wrapped
Specifications
Specifications
Flexible Minitip
Sterile
Sample collection and optimal diagnostics
Nylon Fiber
Individually-wrapped
Flocked Swab