BD Syringe with Luer-Lok Tips (Without Needle)
Siliconized and assembled with scale and tip shield
Supplier: BD 301031
Alert:
NOTICE: The printing has changed on BD 60 mL syringes to only be graduated to 50 mL now. The syringe itself has not changed. Syringes graduated to 60 mL are no longer available.
Description
Siliconized and assembled with scale and tip shield
- Single-use with plastic barrels, plungers and tips
- Nonsterile syringes may by irradiated, autoclaved, or gas sterilized out of polybag
Specifications
Syringe | |
No Needle Included | |
Luer Lock |
1 mL | |
20 mL | |
Non-sterile |