BD Syringe with Luer-Lok Tips (Without Needle)
Siliconized and assembled with scale and tip shield
$329.05 - $482.27
Specifications
Product Type Syringe
Needle Point Number No Needle Included
Connection Type Luer Lock
Sterility Non-sterile
View More Specs
Alert:
NOTICE: The printing has changed on BD 60 mL syringes to only be graduated to 50 mL now. The syringe itself has not changed. Syringes graduated to 60 mL are no longer available.
Products 5
Catalog Number Mfr. No. Volume (Metric) Graduations Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Graduations Price Quantity  
14-823-16D
View Documents
BD
301027
5 mL 1/5 mL
Case of 1400 Each for $403.20
14-823-16E
View Documents
BD
301029
10 mL 1/5 mL
Case of 850 Each for $482.27
14-823-16J
View Documents
BD
301031
20 mL 1 mL
Case of 325 Each for $414.70
14-823-16K
View Documents
BD
301033
30 mL 1 mL
Case of 225 Each for $329.05
14-820-11
View Documents
BD
301035
50 mL 1 mL
Case of 125 Each for $337.50
Description
Description
Siliconized and assembled with scale and tip shield
  • Single-use with plastic barrels, plungers and tips
  • Nonsterile syringes may by irradiated, autoclaved, or gas sterilized out of polybag
Specifications
Specifications
Syringe
No Needle Included
Luer Lock
Non-sterile
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'