missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BD Vacutainer™ Plastic Blood Collection Tubes with Sodium Citrate Edge
Edge

Venous blood collection tubes for routine coagulation studies

Supplier:  BD 363080

 View more versions of this product

Catalog No. 02-683-171



Edge
Explore available promotions

Description

Description

  • Blood is drawn directly from the vein into the evacuated sterile collection tubes
  • Only one venipuncture is needed to draw multiple samples
  • Tubes can be changed while needle and plastic holder remain in place on patient's arm
  • Closures are color coded

  • Made of polyethylene terephthalate (PET)
  • With 3.2% buffered sodium citrate solution
  • Provide a safer work environment for all personnel by (1) reducing risk of tube breakage during handling; (2) eliminating breakage when tubes are centrifuged at higher speeds; and (3) eliminating breakage or chipping of tube rims when stopper is removed and reinserted
  • In addition to reducing potential injuries, fewer tube breakages result in less workflow interruption and fewer patient redraws, providing smoother day-to-day operation

Hemogard™ Stoppers

  • Safety shield designed for safer tube handling
  • Rubber stopper is recessed inside the plastic shield, so any drops left by a blood collection needle remain isolated from potential contact
  • Shield is designed to protect lab personnel from direct contact with blood on the stopper or around the outer rim of the tube, as well as from blood splattering upon opening the tube

Specifications

Specifications

Draw Tube
Plastic
1.8 mL
13 x 75 mm
Buffered Sodium Citrate, 3.2%;
Hemogard
Light Blue
SDS
Documents

Documents

Promotions

Promotions