missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BD Vacutainer™ Plastic Blood Collection Tubes with Sodium Citrate

Venous blood collection tubes for routine coagulation studies

$50.96 - $56.25

Specifications

Material Plastic
Closure Type Hemogard
Diameter (Metric) Outer 13 x 75 mm
View More Specs

Products 2
Catalog Number Mfr. No. Closure Color Capacity (Metric) Product Type Price Quantity  
Catalog Number Mfr. No. Closure Color Capacity (Metric) Product Type Price Quantity  
02-683-172 Edge
View Documents
BD
363083
Blue 2.7 mL Draw Tube
Pack of 100 Each for $50.96
Case of 10 Pack for $509.60
Edge
 
02-683-171 Edge
View Documents
BD
363080
Light Blue 1.8 mL Draw Tube
Pack of 100 Each for $56.25
Case of 10 Pack for $562.50
Edge
 
Description

Description

  • Blood is drawn directly from the vein into the evacuated sterile collection tubes
  • Only one venipuncture is needed to draw multiple samples
  • Tubes can be changed while needle and plastic holder remain in place on patient's arm
  • Closures are color coded

  • Made of polyethylene terephthalate (PET)
  • With 3.2% buffered sodium citrate solution
  • Provide a safer work environment for all personnel by (1) reducing risk of tube breakage during handling; (2) eliminating breakage when tubes are centrifuged at higher speeds; and (3) eliminating breakage or chipping of tube rims when stopper is removed and reinserted
  • In addition to reducing potential injuries, fewer tube breakages result in less workflow interruption and fewer patient redraws, providing smoother day-to-day operation

Hemogard™ Stoppers

  • Safety shield designed for safer tube handling
  • Rubber stopper is recessed inside the plastic shield, so any drops left by a blood collection needle remain isolated from potential contact
  • Shield is designed to protect lab personnel from direct contact with blood on the stopper or around the outer rim of the tube, as well as from blood splattering upon opening the tube

Specifications

Specifications

Plastic
Hemogard
13 x 75 mm
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'