missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BD Vacutainer™ Urine Collection Kits: Microbiology

Reduce the need to retest, relabel, and/or recollect specimens

Supplier:  BD 364951

 View more versions of this product

Catalog No. 02-683-190


Explore available promotions

Description

Description

  • Closed systems that benefit healthcare workers by reducing their need to come into contact with potentially hazardous specimens
  • Designed to provide safety and satisfaction, as well as consistent, reliable results to ensure proper patient outcomes

Microbiology Products

  • Contain a lyophilized preservative that maintains urine for up to 48 hours at room temperature
  • Help to prevent overgrowth without causing toxicity to existing pathogens
  • Minimum fill line (3mL) ensures proper additive-to-urine ratio

Specifications

Specifications

C&S Tube
Sterile
Evacuated culture and sensitivity tube, preservative
Room temperature
4 mL
Plastic
For microbiology Urine testing
Yes
0.135 oz.
Videos
SDS
Documents

Documents

Promotions

Promotions