missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BD Vacutainer™ Urine Collection Kits: Microbiology

Reduce the need to retest, relabel, and/or recollect specimens

$158.14 - $1201.36

Specifications

Material Plastic
Sterility Sterile
Storage Requirements Room temperature
Volume (English) 0.135 oz.
Volume (Metric) 4 mL
View More Specs

Products 3
Catalog Number Mfr. No. Includes Product Type Price Quantity  
Catalog Number Mfr. No. Includes Product Type Price Quantity  
02-683-190
View Documents
BD
364951
Evacuated culture and sensitivity tube, preservative C&S Tube
Case of 1000 Each for $1,201.36
 
02-683-191
View Documents
BD
364953
Preservative and transfer device C&S Transfer Straw Kit
Case of 200 Each for $807.91
 
02-683-192
View Documents
BD
364954
Plastic C and S preservative and transfer straw C&S Cup Kit
Case of 50 Each for $158.14
 
Description

Description

  • Closed systems that benefit healthcare workers by reducing their need to come into contact with potentially hazardous specimens
  • Designed to provide safety and satisfaction, as well as consistent, reliable results to ensure proper patient outcomes

Microbiology Products

  • Contain a lyophilized preservative that maintains urine for up to 48 hours at room temperature
  • Help to prevent overgrowth without causing toxicity to existing pathogens
  • Minimum fill line (3mL) ensures proper additive-to-urine ratio

Specifications

Specifications

Plastic
Room temperature
4 mL
Sterile
0.135 oz.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'