BD Vacutainer™ Urine Collection Kits: Microbiology
Reduce the need to retest, relabel, and/or recollect specimens
Supplier:  BD 364954
 View more versions of this product
Catalog No. 02-683-192
Explore available promotions
Description
Description
  • Closed systems that benefit healthcare workers by reducing their need to come into contact with potentially hazardous specimens
  • Designed to provide safety and satisfaction, as well as consistent, reliable results to ensure proper patient outcomes
Microbiology Products
  • Contain a lyophilized preservative that maintains urine for up to 48 hours at room temperature
  • Help to prevent overgrowth without causing toxicity to existing pathogens
  • Minimum fill line (3mL) ensures proper additive-to-urine ratio
Specifications
Specifications
C&S Cup Kit
Sterile
Plastic C and S preservative and transfer straw
0.135 oz.
Plastic
For Urine testing transfer
Room temperature
4 mL