missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
HemoCue America Beckman Coulter™ Hemoccult™ Fecal Occult Blood Slide Test System
Several packaging configurations are available for different testing needs
Supplier: HemoCue America 60151A
Description
- CLIA Complexity: Waived
- CPT Codes: 82270, G0107
- A convenient, qualitative procedure that uses a guaiac paper slide test to detect fecal occult blood
- On-slide performance monitor enables function and stability of reagents to be tested each time a slide is developed
- Test aids in reducing colorectal cancer with annual screening
- Results in 60 seconds
- Positive results appear as a blue development on the edge of the fecal smear
- Hemoccult II™ Slides feature cards of three tests designed to allow patients to collect three serial specimens at home
- Dispensapak™ and Dispensapak Plus™ Packs are designed for at-home use by patient
- Accessories available; see cat. no. SK63202
Specifications
82270, G0107 | |
Single Slide Kit | |
100 Pack |
Waived | |
100 Single slides, two 15mL bottles of Hemoccult developer, 100 applicator sticks | |
60 sec. |