missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
HemoCue America Beckman Coulter™ Hemoccult™ Fecal Occult Blood Slide Test System
Several packaging configurations are available for different testing needs
$310.84 - $2179.47
Catalog Number | Mfr. No. | Quantity | Includes | Type | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Includes | Type | Price | Quantity | |||
SK-61200
![]() |
HemoCue America
61200A |
102 Pack | 34 Triple slides, 2 x 15mL bottles of Hemoccult developer, 100 applicator sticks Kit | Triple Slide kit |
Pack of 102 Each for $310.84
Case of 10 Pack for $3,108.40
|
|
|||
SK-61130
![]() |
HemoCue America
61130A |
40 Pack | 6 x 15mL Bottles of developer and 40 patient kits (1 triple slide, 3 flushable sample collection tissues, 3 applicator, diet and test instructions and barrier mailing envelope) | Dispensapak™ Plus Kit |
Case of 40 Each for $1,460.87
|
|
|||
SK-60151
![]() |
HemoCue America
60151A |
100 Pack | 100 Single slides, two 15mL bottles of Hemoccult developer, 100 applicator sticks | Single Slide Kit |
Case of 100 Each for $2,179.47
|
|
|||
SK-61100
![]() |
HemoCue America
BEC61100 |
100 Case | 6 x 15mL Bottles of developer and 100 patient kits (1 triple slide, 3 applicator and diet and test instructions) | Dispensapak™ Kit |
Pack of 100 Each for $824.66
|
|
|||
Description
- CLIA Complexity: Waived
- CPT Codes: 82270, G0107
- A convenient, qualitative procedure that uses a guaiac paper slide test to detect fecal occult blood
- On-slide performance monitor enables function and stability of reagents to be tested each time a slide is developed
- Test aids in reducing colorectal cancer with annual screening
- Results in 60 seconds
- Positive results appear as a blue development on the edge of the fecal smear
- Hemoccult II™ Slides feature cards of three tests designed to allow patients to collect three serial specimens at home
- Dispensapak™ and Dispensapak Plus™ Packs are designed for at-home use by patient
- Accessories available; see cat. no. SK63202
Specifications
82270, G0107 | |
Waived | |
60 sec. |