HemoCue America Beckman Coulter™ Hemoccult™ Fecal Occult Blood Slide Test System DFS Item
Several packaging configurations are available for different testing needs
Supplier:  HemoCue America BEC61100
 View more versions of this product
Catalog No. SK-61100
Explore available promotions
Description
Description
  • CLIA Complexity: Waived
  • CPT Codes: 82270, G0107
  • A convenient, qualitative procedure that uses a guaiac paper slide test to detect fecal occult blood
  • On-slide performance monitor enables function and stability of reagents to be tested each time a slide is developed
  • Test aids in reducing colorectal cancer with annual screening
  • Results in 60 seconds
  • Positive results appear as a blue development on the edge of the fecal smear
  • Hemoccult II™ Slides feature cards of three tests designed to allow patients to collect three serial specimens at home
  • Dispensapak™ and Dispensapak Plus™ Packs are designed for at-home use by patient
  • Accessories available; see cat. no. SK63202
Specifications
Specifications
82270, G0107
Dispensapak™ Kit
100 Case
Waived
6 x 15mL Bottles of developer and 100 patient kits (1 triple slide, 3 applicator and diet and test instructions)
60 sec.