missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bel-Art™ SP Scienceware™ Clear View Fume Hood
Safely vent noxious or potentially hazardous fumes and dust with the SP Scienceware™ Clear View Fume Hood.
Supplier: Bel-Art™ F500202010
Includes: Laboratory, classroom or industrial
Description
- Versatile 3-position sash
- Clear polycarbonate provides optimum visibility and durability
500201010
- Top sitting: 6 in. O.D. vent
- O.D.: 24D x 25.8W x 26.12 in. H (61 x 65.7 x 66.4cm)
- I.D.: 20D x 24W x 24 in. H (50.8 x 61 x 61cm)
- Internal Space: 11.67 cu. ft. (0.18m)
- Two Top sitting: 6 in. O.D. vent
- O.D.: 24D x 43.8W x 26.1 in. H (61 x 111.4 x 66.4cm)
- I.D.: 20D x 42W x 24 in. H (50.8 x 106.7 x 61cm)
- Internal Space: 11.67 cu. ft. (0.18m)
Specifications
Fume Hood | |
24 in. | |
26 in. | |
111.4 cm |
Polycarbonate | |
61 cm | |
43.87 in. |