missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bel-Art™ SP Scienceware™ Clear View Fume Hood
Safely vent noxious or potentially hazardous fumes and dust with the SP Scienceware™ Clear View Fume Hood.
$4276.26 - $5013.21
Specifications
Product Type | Fume Hood |
---|---|
Material | Polycarbonate |
Depth (English) | 24 in. |
Depth (Metric) | 61 cm |
Height (English) | 26 in. |
Includes: Laboratory, classroom or industrial
Catalog Number | Mfr. No. | Width (English) | Width (Metric) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Width (English) | Width (Metric) | Price | Quantity | ||||
03-421-123
|
Bel-Art™
F500201010 |
25.87 in. | 65.7 cm |
Each for $4,276.26
|
|
||||
03-421-124
|
Bel-Art™
F500202010 |
43.87 in. | 111.4 cm |
Each for $5,013.21
|
|
||||
Description
- Versatile 3-position sash
- Clear polycarbonate provides optimum visibility and durability
500201010
- Top sitting: 6 in. O.D. vent
- O.D.: 24D x 25.8W x 26.12 in. H (61 x 65.7 x 66.4cm)
- I.D.: 20D x 24W x 24 in. H (50.8 x 61 x 61cm)
- Internal Space: 11.67 cu. ft. (0.18m)
- Two Top sitting: 6 in. O.D. vent
- O.D.: 24D x 43.8W x 26.1 in. H (61 x 111.4 x 66.4cm)
- I.D.: 20D x 42W x 24 in. H (50.8 x 106.7 x 61cm)
- Internal Space: 11.67 cu. ft. (0.18m)
Specifications
Fume Hood | |
24 in. | |
26 in. |
Polycarbonate | |
61 cm |