Bel-Art™ SP Scienceware™ Double Spinfin™ Magnetic Stirring Bars
To agitate liquids in round bottom vessels where other stirring bars cannot be used.
Supplier:  Bel-Art™ F371260034
 View more versions of this product
Catalog No. 14-512-173
Explore available promotions
Description
Description
  • Made from PTFE
  • Fins on top and bottom ensure complete agitation
  • Does not wobble, maintains position in vessel
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE low friction coating
  • Durable and inert
  • Autoclavable
Specifications
Specifications
19 mm
16mm
50
Alnico Magnet
Cylinder
0.75 in.
White
0.62 in.
FDA