Bel-Art™ SP Scienceware™ Double Spinfin™ Magnetic Stirring Bars
To agitate liquids in round bottom vessels where other stirring bars cannot be used.
$36.46 - $66.28
Catalog Number | Mfr. No. | Diameter (English) | Diameter (Metric) | Height (English) | Height (Metric) | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (English) | Diameter (Metric) | Height (English) | Height (Metric) | Price | Quantity | ||
14-512-172
|
Bel-Art™
F371260012 |
0.5 in. | 12.7 mm | 0.5 in. | 12.7mm |
Each for $56.10
Case of 50 Each for $2,805.00
|
|
||
14-512-173
|
Bel-Art™
F371260034 |
0.75 in. | 19 mm | 0.62 in. | 16mm |
Each for $66.28
Case of 50 Each for $3,314.00
|
|
||
14-512-174
|
Bel-Art™
F371260001 |
1 in. | 25.4 mm | 0.75 in. | 19.1mm |
Each for $36.46
Case of 50 Each for $1,823.00
|
|
||
Description
- Made from PTFE
- Fins on top and bottom ensure complete agitation
- Does not wobble, maintains position in vessel
- Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE low friction coating
- Durable and inert
- Autoclavable