Bel-Art™ Double Spinfin™ Magnetic Stirring Bar
To agitate liquids in round bottom vessels where other stirring bars cannot be used.
Supplier:  Bel-Art™ F371260012
 View more versions of this product
Catalog No. 14-512-172
Explore available promotions
Description
Description
  • Made from PTFE
  • Fins on top and bottom ensure complete agitation
  • Does not wobble, maintains position in vessel
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE low friction coating
  • Durable and inert
  • Autoclavable
Specifications
Specifications
12.7 mm
0.5 in.
Stir Bar
0.5 in.
12.7 mm
PTFE, FDA-grade, USP Class VI
12.7mm
1
PTFE
Double Crosshead