Bel-Art™ Dry-Keeper™ Desiccator Cabinet
Clear, durable polystyrene desiccator cabinets have black ABS accents and movable shelves for adjusting dry storage space configuration for moisture-sensitive materials.
$3119.71 - $4248.33
Specifications
Material Methyl Methacrylate
Product Type Desiccator
Door Style Hinged and gasketed
For Use With (Application) Adjusting dry storage space configuration for moisture-sensitive materials, can be used with Scienceware™ desiccant cartridges, canisters or similar desiccant
Volume (English) 2 cu. ft.
View More Specs
Products 2
Catalog Number Mfr. No. Color Cabinet Style No. of Shelves Holds Price Quantity  
Catalog Number Mfr. No. Color Cabinet Style No. of Shelves Holds Price Quantity  
08-647-30
View Documents
Bel-Art™
H420580003
Clear Horizontal 2 Adjustable 87 Cans
Each for $4,248.33
08-647-28
View Documents
Bel-Art™
H420580001
Translucent Horizontal 2 adjustable perforated shelves 78 Cans
Each for $3,119.71
Description
Description
  • Generous 2 cu. ft. interior volume
  • Adjustable, removable, and perforated shelves facilitate full circulation of air; additional shelves and rails sold separately
  • Large, in-door dial hygrometer for easy visualization of RH%
  • Gasketed door closes with two latches for a tight seal
  • Standard cabinets include desiccant tray (desiccant sold separately)
Horizontal Cabinets
  • Include two shelves
  • Shelf rails can be repositioned on any one of eight steps set at 1.1 in. (28 mm) intervals
  • Interior opening measures 17.75 x 9 in. (45 x 23 cm)
Vertical Cabinets
  • Includes three shelves
  • Shelf rails can be repositioned on any one of 15 steps set at 1.1 in. (28 mm) intervals
  • Interior opening measures 10 x 17 in. (25 x 43 cm)
Autodesiccator (H42058-0003, H42056-1003)
  • 120VAC electronic dehumidification system maintains relative humidity levels at 30 to 40%†; keeps dry air circulating throughout the cabinet
  • Permanent desiccant contained in the control unit
  • Red light indicates operation of the 4.5 hour absorption period; green light indicates operation of the 0.5 hour regeneration of the desiccant
†Cabinet interior RH% levels can differ depending upon room temperature, room RH%, and usage conditions.
Specifications
Specifications
Methyl Methacrylate
Hinged and gasketed
2 cu. ft.
Desiccator
Adjusting dry storage space configuration for moisture-sensitive materials, can be used with Scienceware™ desiccant cartridges, canisters or similar desiccant
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'