Bel-Art™ SP Scienceware™ Lab Companion Round Style Vacuum Desiccators
Easy to view vacuum gauge eliminates the guesswork
Supplier:  Bel-Art™ F424002241
 View more versions of this product
Catalog No. 08-648-105
Explore available promotions
Includes: Desiccant tray (desiccants sold separately)
Description
Description
High-quality vacuum desiccators provide vacuum integrity lasting three times longer than the average vacuum desiccator resulting in lower maintenance and greater peace of mind. Able to maintain 29in.-Hg vacuum for over 72 hours at room temperature, they stay gastight allowing reliable storage or experiments over extended periods. A Viton™ 3-way valve offers consistent and uniform vacuum draw, vacuum release, or gas exchange. Transparent polycarbonate construction is durable, easy-to-clean, chemical-resistant, and allows quick visualization of the desiccator contents. Amber tinted UV blocking models block 100% of UVA, B, and C light to minimize damages or discoloration of light-sensitive samples. Use with any vacuum with free air capacity of 1.5 cubic feet per minute (CFM) ft or greater.
  • Vacuum level checking is simplified with a built-in analog vacuum gauge at the top of the unit
  • High-quality silicone O-ring and specially designed locking ring allow outstanding vacuum capability with virtually no leakage
  • Locking ring secures top and bottom halves when vacuum not in use; provides additional convenience and safety when moving the unit
  • Six models to choose from; 6, 10, and 20 liter internal volume in clear or UV blocking amber
Specifications
Specifications
UV Amber
Desiccant tray, built-in analog vacuum gauge, silicone o-ring and locking ring, Viton™ 3-way valve
1
0.7 cu. ft.
16.75 in.
47.5 cm
9.5 lb.
Vacuum Dessicator
25 kg
Round
20 L
42.7 cm
18.75 in.
4.3 kg