Bel-Art™ SP Scienceware™ OdoClave™ Deodorant Autoclave Pads
Neutralize autoclave odors with a pleasant fragrance
Supplier:  Bel-Art™ H131980001
 View more versions of this product
Catalog No. 14-488-32
Explore available promotions
Description
Description
Neutralize autoclave odors with a pleasant fragrance
  • Heat-release deodorant works automatically at autoclave temperatures
  • Open pouch and place in bag prior to autoclaving
  • Three-year shelf life or longer if stored in a sealed container
  • 32mm (1.75 in.) square
  • Triple strength pads release three times the deodorant of standard pads while costing less than two standard pads
Order Info
Choose 1.25 in. (32mm) square regular-strength or 2.25 in. (5.7cm) square super-strength pad.
Specifications
Specifications
Cherry
Autoclavable
Red