Bel-Art™ Polypropylene Graduated Pitchers
Constructed of translucent polypropylene. SP Scienceware™ Polypropylene Graduated Pitchers are ideal for general laboratory use and offer well designed spouts and large handles.
$22.26 - $502.03
Specifications
Material Polypropylene
For Use With (Application) For Controlled Pouring, Chemically Resistant and Shatterproof
Temperature (English) Max. Operating 250°F
Temperature (Metric) Max. Operating 121°C
Type Pitcher
View More Specs
Products 7
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
22-163683
View Documents
Bel-Art™
F289890000
250 mL
Case of 24 for $502.03
22-163691
View Documents
Bel-Art™
F289900000
500 mL
Each for $22.26
Case of 24 Each for $534.24
22-163709
View Documents
Bel-Art™
F289910000
1000 mL
Each for $31.24
Case of 24 Each for $749.76
22-163717
View Documents
Bel-Art™
F289920000
2000 mL
Each for $35.32
Case of 12 Each for $423.84
22-171-124
View Documents
Bel-Art™
F289930000
3000 mL
Each for $49.32
Case of 12 Each for $591.84
22-171132
View Documents
Bel-Art™
F289940000
5000 mL
Each for $118.53
Case of 6 Each for $711.18
14-513-304
View Documents
Bel-Art™
F289950000
10 L
Each for $356.09
Description
Description
Constructed of translucent polypropylene. SP Scienceware™ Polypropylene Graduated Pitchers are ideal for general laboratory use and offer well designed spouts and large handles.
  • Metric graduations molded into outside wall
  • Steam autoclavable at 121°C (250°F)
  • Balanced handle for controlled pouring
  • 5000mL (5L) and 10000mL (10L) models feature an updated molded handle for a more secure grip, and a redesigned hand-slot at base for easy lifting
Specifications
Specifications
Polypropylene
250°F
Pitcher
Yes
For Controlled Pouring, Chemically Resistant and Shatterproof
121°C
Autoclavable
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'