missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bel-Art™ SP Scienceware™ Red Wash Bottles
The bright red color helps identify hazardous contents at a glance
Supplier: Bel-Art™ F116500016
Description
- Low-density polyethylene with polypropylene closure
Specifications
16 oz., 500 mL | |
16 oz. | |
LDPE | |
Narrow | |
28 mm |
500 mL | |
Red | |
Not Autoclavable | |
Polypropylene |