Bel-Art™ SP Scienceware™ Write-On™ Multi-Roll Label Tape Dispensers
Provide smooth writing surface for multiple tapes of different colors and width, making labeling easy and fast
Supplier:  Bel-Art™ F134730000
 View more versions of this product
Catalog No. 03-410-489
Explore available promotions
Description
Description
  • Various width and color tape rolls can be mixed together on spindles, which accommodate 5 or more 0.5 to 0.75 in. (12 to 19mm) narrow rolls or four 1 in. (25mm) wide rolls
  • When marked tape is advanced and cut off on serrated edge, a fresh length of label tape is positioned on writing platform
  • Spindles are easily removed from unit for loading and unloading, and can be adjusted to hold either 1 or 3 in. (25 or 75mm) core diameter rolls
  • Dispenser includes three plastic separating discs to assure smooth, snag free dispensing for up to four rolls of tape
  • Molded-in pen holder wells on the dispenser help to keep writing instruments handy
  • 12 × 5.63 × 4.13 in. (302 × 143 × 121mm)
  • Available in choice of economy and benchtop models
  • Additional separating discs are available separately
Benchtop Model
  • Plastic coated, weighted steel base with rubber feet to prevent slippage
  • Steel serrated cutting edge for long term use
  • Includes one free roll of 1/2™, White, Write-On Labeling Tape
Economy Model:
  • Designed for lighter duty use as the plastic cutting edge is molded into housing
  • Includes mounting holes for secure attachment to a benchtop (screws not supplied)
  • Can be attached to vertical surfaces using Wall Mount adapter
  • Includes one free roll of 1/2™, White, Write-On Labeling Tape
Specifications
Specifications
Label Tape Dispenser
Provides a smooth writing surface, making labeling easy and fast, molded-in pen holder wells on the dispenser help to keep writing instruments handy
Three plastic separating discs to assure smooth, snag free dispensing for up to four rolls of tape, one free roll of 0.5 in. white Write-On™ labeling tape
Tapes with 25 mm or 75 mm Core
Full line of 13463-series Write-On™ Tapes, which are available in 6 colors and 3 wide
Economy