Fisherbrand™ Bench Protector
Fisherbrand™ Bench Protector features absorbent paper to capture spills, backed with an impermeable polyethylene to protect the covered surface from contamination
Supplier:  Fisherbrand™ 948844
 View more versions of this product
Catalog No. FB12007300
Explore available promotions
Description
Description
Features:
  • Absorbent layer of pure cellulose with smooth, firm surface
  • One side coated with impermeable polyethylene
  • Dry and liquid samples can be recovered
  • Can be incinerated for disposal
Specifications
Specifications
Bench Protector
25 mg/cm2
57 cm
46 cm
White
Single-use
50
Cellulose/Polyethylene
22.44 in.
18.11 in.
46 x 57 cm
Sheet
Protects benchtops, floors and carts with this highly absorbent cover
Pk. of 50