Fisherbrand™ Infrared Micro-Sterilizer Accessories
Designed for use with Bacti-Loop™ micro sterilizer.
Supplier:  Fisherbrand™ B100001
 View more versions of this product
Catalog No. 22-630-002
Explore available promotions
Description
Description
Loop Holder Attachment for use with Fisher HealthCare™ Bacti-Loop™ Infrared Micro-Sterilizer
Specifications
Specifications
15 cm
Ideal for platinum loops, needles and tube mouths
6.5 in.
Loop Holder Attachment
Stainless Steel
120 V
10 cm
1.39 cm
Bacti-Loop Micro-Sterilizer
16.5 cm
50/60 Hz
5 to 7 sec.
4 in.