missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzaldehyde Dimethyl Acetal 98.0+%, TCI America™

Supplier:  TCI America B1197500ML

 View more versions of this product

Catalog No. B1197500ML

Explore available promotions

Description

Description

Specifications

Specifications

Benzaldehyde Dimethyl Acetal
Colorless
C9H12O2
500 mL
HEVMDQBCAHEHDY-UHFFFAOYSA-N
dimethoxymethylbenzene
62375
≥98.0% (GC)
1125-88-8
196°C
MFCD00008491
benzaldehyde dimethyl acetal, dimethoxymethyl benzene, alpha,alpha-dimethoxytoluene, dimethoxyphenylmethane, benzene, dimethoxymethyl, benzaldehydedimethylacetal, dimethoxymethyl-benzene, benzaldehyde, dimethyl acetal, benzaldehyde dimethylacetal, unii-w6t83k35ca
COC(C1=CC=CC=C1)OC
152.193
152.19
Liquid

Chemical Identifiers

1125-88-8
152.193
HEVMDQBCAHEHDY-UHFFFAOYSA-N
62375
COC(C1=CC=CC=C1)OC
C9H12O2
MFCD00008491
benzaldehyde dimethyl acetal, dimethoxymethyl benzene, alpha,alpha-dimethoxytoluene, dimethoxyphenylmethane, benzene, dimethoxymethyl, benzaldehydedimethylacetal, dimethoxymethyl-benzene, benzaldehyde, dimethyl acetal, benzaldehyde dimethylacetal, unii-w6t83k35ca
dimethoxymethylbenzene
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1152

RTECSNumber : CU5774000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions