missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzaldehyde Dimethyl Acetal 98.0+%, TCI America™
$48.46 - $292.65
Chemical Identifiers
| CAS | 1125-88-8 |
|---|---|
| Molecular Formula | C9H12O2 |
| Molecular Weight (g/mol) | 152.193 |
| MDL Number | MFCD00008491 |
| InChI Key | HEVMDQBCAHEHDY-UHFFFAOYSA-N |
| Synonym | benzaldehyde dimethyl acetal, dimethoxymethyl benzene, alpha,alpha-dimethoxytoluene, dimethoxyphenylmethane, benzene, dimethoxymethyl, benzaldehydedimethylacetal, dimethoxymethyl-benzene, benzaldehyde, dimethyl acetal, benzaldehyde dimethylacetal, unii-w6t83k35ca |
| PubChem CID | 62375 |
| IUPAC Name | dimethoxymethylbenzene |
| SMILES | COC(C1=CC=CC=C1)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B119725ML
|
TCI America
B119725ML |
25 mL |
Each for $48.46
|
|
|||||
|
B1197100ML
|
TCI America
B1197100ML |
100 mL |
Each for $89.48
|
|
|||||
|
B1197500ML
|
TCI America
B1197500ML |
500 mL |
Each for $292.65
|
|
|||||
Chemical Identifiers
| 1125-88-8 | |
| 152.193 | |
| HEVMDQBCAHEHDY-UHFFFAOYSA-N | |
| 62375 | |
| COC(C1=CC=CC=C1)OC |
| C9H12O2 | |
| MFCD00008491 | |
| benzaldehyde dimethyl acetal, dimethoxymethyl benzene, alpha,alpha-dimethoxytoluene, dimethoxyphenylmethane, benzene, dimethoxymethyl, benzaldehydedimethylacetal, dimethoxymethyl-benzene, benzaldehyde, dimethyl acetal, benzaldehyde dimethylacetal, unii-w6t83k35ca | |
| dimethoxymethylbenzene |
Specifications
| 1125-88-8 | |
| 196°C | |
| MFCD00008491 | |
| benzaldehyde dimethyl acetal, dimethoxymethyl benzene, alpha,alpha-dimethoxytoluene, dimethoxyphenylmethane, benzene, dimethoxymethyl, benzaldehydedimethylacetal, dimethoxymethyl-benzene, benzaldehyde, dimethyl acetal, benzaldehyde dimethylacetal, unii-w6t83k35ca | |
| COC(C1=CC=CC=C1)OC | |
| 152.193 | |
| 152.19 | |
| Liquid |
| Colorless | |
| C9H12O2 | |
| 25 mL | |
| HEVMDQBCAHEHDY-UHFFFAOYSA-N | |
| dimethoxymethylbenzene | |
| 62375 | |
| ≥98.0% (GC) | |
| Benzaldehyde Dimethyl Acetal |
Safety and Handling
EINECSNumber : (3)-1152
RTECSNumber : CU5774000
TSCA : Yes