Learn More
Benzamidine Hydrochloride Hydrate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 105241000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Benzamidine hydrochloride hydrate | |
97.5 | |
80°C to 86°C | |
Authentic | |
Plastic bottle | |
HCl·xH2O | |
100 g | |
Solubility in water: soluble | |
Cl.NC(=N)C1=CC=CC=C1 | |
156.61 | |
156.61 | |
Powder |
206752-36-5,1670-14-0 | |
100.0 | |
White | |
97.5 to 102.5% (on dry substance) (Argentometry) | |
C7H9ClN2 | |
MFCD00066285 | |
benzamidine hydrochloride hydrate, benzenecarboximidamide hydrochloride hydrate, benzamidine hydrate hydrochloride, benzamidinium chloride, benzamidinehydrochloridehydrate, amidinobenzene hydrochloride, benzamidine hydrochloride monohydrate, acmc-20a6bw, ksc491m3d, benzamidin hydrochloride hydrate | |
LZCZIHQBSCVGRD-UHFFFAOYSA-N | |
benzenecarboximidamide;hydrate;hydrochloride | |
16219042 | |
98% |
Chemical Identifiers
206752-36-5 | |
156.61 | |
LZCZIHQBSCVGRD-UHFFFAOYSA-N | |
16219042 | |
Cl.NC(=N)C1=CC=CC=C1 |
C7H9ClN2 | |
MFCD00066285 | |
benzamidine hydrochloride hydrate, benzenecarboximidamide hydrochloride hydrate, benzamidine hydrate hydrochloride, benzamidinium chloride, benzamidinehydrochloridehydrate, amidinobenzene hydrochloride, benzamidine hydrochloride monohydrate, acmc-20a6bw, ksc491m3d, benzamidin hydrochloride hydrate | |
benzenecarboximidamide;hydrate;hydrochloride |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/
GHS Signal Word: Warning
RUO – Research Use Only