missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzamidine Hydrochloride Hydrate, 98%, Thermo Scientific Chemicals

$52.88 - $1355.12

Chemical Identifiers

CAS 206752-36-5
Molecular Formula C7H9ClN2
Molecular Weight (g/mol) 156.61
MDL Number MFCD00066285
InChI Key LZCZIHQBSCVGRD-UHFFFAOYSA-N
Synonym benzamidine hydrochloride hydrate, benzenecarboximidamide hydrochloride hydrate, benzamidine hydrate hydrochloride, benzamidinium chloride, benzamidinehydrochloridehydrate, amidinobenzene hydrochloride, benzamidine hydrochloride monohydrate, acmc-20a6bw, ksc491m3d, benzamidin hydrochloride hydrate
PubChem CID 16219042
IUPAC Name benzenecarboximidamide;hydrate;hydrochloride
SMILES Cl.NC(=N)C1=CC=CC=C1
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC105240050
SDS View Documents
Thermo Scientific Chemicals
105240050
5 g Glass Bottle
Each for $52.88
 
AC105240250
SDS View Documents
Thermo Scientific Chemicals
105240250
25 g Plastic bottle
Each for $148.51
 
AC105241000
SDS View Documents
Thermo Scientific Chemicals
105241000
100 g Plastic bottle
Each for $417.81
 
AC105245000
SDS View Documents
Thermo Scientific Chemicals
105245000
500 g Plastic bottle
Each for $1,355.12
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

206752-36-5
156.61
LZCZIHQBSCVGRD-UHFFFAOYSA-N
16219042
Cl.NC(=N)C1=CC=CC=C1
C7H9ClN2
MFCD00066285
benzamidine hydrochloride hydrate, benzenecarboximidamide hydrochloride hydrate, benzamidine hydrate hydrochloride, benzamidinium chloride, benzamidinehydrochloridehydrate, amidinobenzene hydrochloride, benzamidine hydrochloride monohydrate, acmc-20a6bw, ksc491m3d, benzamidin hydrochloride hydrate
benzenecarboximidamide;hydrate;hydrochloride

Specifications

206752-36-5,1670-14-0
100.0
White
97.5 to 102.5% (on dry substance) (Argentometry)
C7H9ClN2
MFCD00066285
benzamidine hydrochloride hydrate, benzenecarboximidamide hydrochloride hydrate, benzamidine hydrate hydrochloride, benzamidinium chloride, benzamidinehydrochloridehydrate, amidinobenzene hydrochloride, benzamidine hydrochloride monohydrate, acmc-20a6bw, ksc491m3d, benzamidin hydrochloride hydrate
LZCZIHQBSCVGRD-UHFFFAOYSA-N
benzenecarboximidamide;hydrate;hydrochloride
16219042
98%
Benzamidine hydrochloride hydrate
97.5
80°C to 86°C
Authentic
Glass Bottle
HCl·xH2O
5 g
Solubility in water: soluble
Cl.NC(=N)C1=CC=CC=C1
156.61
156.61
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only