missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzamidine Hydrochloride Hydrate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 105245000

 View more versions of this product

Catalog No. AC105245000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Benzamidine hydrochloride hydrate
97.5
80°C to 86°C
Authentic
Plastic bottle
HCl·xH2O
500 g
Solubility in water: soluble
Cl.NC(=N)C1=CC=CC=C1
156.61
156.61
Powder
206752-36-5,1670-14-0
100.0
White
97.5 to 102.5% (on dry substance) (Argentometry)
C7H9ClN2
MFCD00066285
benzamidine hydrochloride hydrate, benzenecarboximidamide hydrochloride hydrate, benzamidine hydrate hydrochloride, benzamidinium chloride, benzamidinehydrochloridehydrate, amidinobenzene hydrochloride, benzamidine hydrochloride monohydrate, acmc-20a6bw, ksc491m3d, benzamidin hydrochloride hydrate
LZCZIHQBSCVGRD-UHFFFAOYSA-N
benzenecarboximidamide;hydrate;hydrochloride
16219042
98%

Chemical Identifiers

206752-36-5
156.61
LZCZIHQBSCVGRD-UHFFFAOYSA-N
16219042
Cl.NC(=N)C1=CC=CC=C1
C7H9ClN2
MFCD00066285
benzamidine hydrochloride hydrate, benzenecarboximidamide hydrochloride hydrate, benzamidine hydrate hydrochloride, benzamidinium chloride, benzamidinehydrochloridehydrate, amidinobenzene hydrochloride, benzamidine hydrochloride monohydrate, acmc-20a6bw, ksc491m3d, benzamidin hydrochloride hydrate
benzenecarboximidamide;hydrate;hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only