missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzeneseleninic anhydride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 199710050

 View more versions of this product

Catalog No. AC199710050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Benzeneseleninic anhydride
97.5
170°C to 173°C
Authentic
Glass bottle
[C6H5Se(O)]2O
5 g
06,240; 07,139; 08,29; 09,32; 10,22; 11,37; 15,18
FHPZOWOEILXXBD-UHFFFAOYSA-N
phenylseleninyl benzeneseleninate
87253
98%
17697-12-0
100.0
White to Beige
98%
C12H10O3Se2
MFCD00001991
3283
benzeneseleninic anhydride, benzeneseleninic acid anhydride, benzeneseleninyl benzeneseleninate, bis phenylseleninic anhydride, benzeneseleninic acid, anhydride, bis benzeneseleninic anhydride, 1,3-diphenyldiselenoxane 1,3-dioxide #
C1=CC=C(C=C1)[Se](=O)O[Se](=O)C2=CC=CC=C2
360.13
360.13
Powder

Chemical Identifiers

17697-12-0
360.13
FHPZOWOEILXXBD-UHFFFAOYSA-N
87253
C1=CC=C(C=C1)[Se](=O)O[Se](=O)C2=CC=CC=C2
C12H10O3Se2
MFCD00001991
benzeneseleninic anhydride, benzeneseleninic acid anhydride, benzeneseleninyl benzeneseleninate, bis phenylseleninic anhydride, benzeneseleninic acid, anhydride, bis benzeneseleninic anhydride, 1,3-diphenyldiselenoxane 1,3-dioxide #
phenylseleninyl benzeneseleninate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.
Toxic if inhaled.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge

GHS Signal Word: Danger

EINECSNumber : 241-701-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only