missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzeneseleninic anhydride, 98%, Thermo Scientific Chemicals

$226.68 - $752.99

Chemical Identifiers

CAS 17697-12-0
Molecular Formula C12H10O3Se2
Molecular Weight (g/mol) 360.13
MDL Number MFCD00001991
InChI Key FHPZOWOEILXXBD-UHFFFAOYSA-N
Synonym benzeneseleninic anhydride, benzeneseleninic acid anhydride, benzeneseleninyl benzeneseleninate, bis phenylseleninic anhydride, benzeneseleninic acid, anhydride, bis benzeneseleninic anhydride, 1,3-diphenyldiselenoxane 1,3-dioxide #
PubChem CID 87253
IUPAC Name phenylseleninyl benzeneseleninate
SMILES C1=CC=C(C=C1)[Se](=O)O[Se](=O)C2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC199710050
SDS View Documents
Thermo Scientific Chemicals
199710050
5 g Glass bottle
Each for $226.68
 
AC199710250
SDS View Documents
Thermo Scientific Chemicals
199710250
25 g Glass bottle
Each for $752.99
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

17697-12-0
360.13
FHPZOWOEILXXBD-UHFFFAOYSA-N
87253
C1=CC=C(C=C1)[Se](=O)O[Se](=O)C2=CC=CC=C2
C12H10O3Se2
MFCD00001991
benzeneseleninic anhydride, benzeneseleninic acid anhydride, benzeneseleninyl benzeneseleninate, bis phenylseleninic anhydride, benzeneseleninic acid, anhydride, bis benzeneseleninic anhydride, 1,3-diphenyldiselenoxane 1,3-dioxide #
phenylseleninyl benzeneseleninate

Specifications

17697-12-0
100.0
White to Beige
98%
C12H10O3Se2
MFCD00001991
3283
benzeneseleninic anhydride, benzeneseleninic acid anhydride, benzeneseleninyl benzeneseleninate, bis phenylseleninic anhydride, benzeneseleninic acid, anhydride, bis benzeneseleninic anhydride, 1,3-diphenyldiselenoxane 1,3-dioxide #
C1=CC=C(C=C1)[Se](=O)O[Se](=O)C2=CC=CC=C2
360.13
360.13
Powder
97.5
170°C to 173°C
Authentic
Glass bottle
[C6H5Se(O)]2O
5 g
06,240; 07,139; 08,29; 09,32; 10,22; 11,37; 15,18
FHPZOWOEILXXBD-UHFFFAOYSA-N
phenylseleninyl benzeneseleninate
87253
98%
Benzeneseleninic anhydride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.
Toxic if inhaled.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge

GHS Signal Word: Danger

EINECSNumber : 241-701-3

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only