missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzo[a]phenanthrene (purified by sublimation) 98.0+%, TCI America™

$162.63 - $487.89

Chemical Identifiers

CAS 218-01-9
Molecular Formula C18H12
Molecular Weight (g/mol) 228.294
MDL Number MFCD00003698
InChI Key WDECIBYCCFPHNR-UHFFFAOYSA-N
Synonym benzo a phenanthrene, 1,2-benzophenanthrene, 1,2-benzphenanthrene, 1,2,5,6-dibenzonaphthalene, benz a phenanthrene, chrysen, rcra waste number u050, rcra waste no. u050
PubChem CID 9171
ChEBI CHEBI:51687
IUPAC Name chrysene
SMILES C1=CC=C2C(=C1)C=CC3=C2C=CC4=CC=CC=C43
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C0339100MG
View Documents
TCI America
C0339100MG
100 mg
Each for $162.63
 
C03391G
View Documents
TCI America
C03391G
1 g
Each for $487.89
 
Description

Description

Specifications

Chemical Identifiers

218-01-9
228.294
WDECIBYCCFPHNR-UHFFFAOYSA-N
9171
chrysene
C18H12
MFCD00003698
benzo a phenanthrene, 1,2-benzophenanthrene, 1,2-benzphenanthrene, 1,2,5,6-dibenzonaphthalene, benz a phenanthrene, chrysen, rcra waste number u050, rcra waste no. u050
CHEBI:51687
C1=CC=C2C(=C1)C=CC3=C2C=CC4=CC=CC=C43

Specifications

218-01-9
448°C
MFCD00003698
3077
WDECIBYCCFPHNR-UHFFFAOYSA-N
chrysene
9171
228.29
Crystalline Powder
255°C
C18H12
100 mg
benzo a phenanthrene, 1,2-benzophenanthrene, 1,2-benzphenanthrene, 1,2,5,6-dibenzonaphthalene, benz a phenanthrene, chrysen, rcra waste number u050, rcra waste no. u050
C1=CC=C2C(=C1)C=CC3=C2C=CC4=CC=CC=C43
228.294
CHEBI:51687
≥98.0% (GC)
Benzo[a]phenanthrene (purified by sublimation)
Videos
Safety and Handling

Safety and Handling

RTECSNumber : GC0700000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'