missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Benzo[ghi]perylene, 98+%, Thermo Scientific Chemicals
$202.72 - $590.35
Chemical Identifiers
CAS | 191-24-2 |
---|---|
Molecular Formula | C22H12 |
Molecular Weight (g/mol) | 276.33 |
MDL Number | MFCD00004135 |
InChI Key | GYFAGKUZYNFMBN-UHFFFAOYSA-N |
Synonym | benzo ghi perylene, 1,12-benzoperylene, 1,12-benzperylene, benzo g,h,i perylene, benzo ghi pyrilene, benzo-1,12-perylene, unii-5zme2e2q9l, ccris 784 |
PubChem CID | 9117 |
ChEBI | CHEBI:34568 |
SMILES | C1=CC2=C3C(=C1)C4=CC=CC5=C4C6=C(C=C5)C=CC(=C36)C=C2 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC105550250
|
Thermo Scientific Chemicals
105550250 |
25 mg | Glass bottle |
Each for $202.72
|
|
||||
AC105551000
|
Thermo Scientific Chemicals
105551000 |
100 mg | Glass bottle |
Each for $590.35
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
191-24-2 | |
276.33 | |
GYFAGKUZYNFMBN-UHFFFAOYSA-N | |
9117 | |
C1=CC2=C3C(=C1)C4=CC=CC5=C4C6=C(C=C5)C=CC(=C36)C=C2 |
C22H12 | |
MFCD00004135 | |
benzo ghi perylene, 1,12-benzoperylene, 1,12-benzperylene, benzo g,h,i perylene, benzo ghi pyrilene, benzo-1,12-perylene, unii-5zme2e2q9l, ccris 784 | |
CHEBI:34568 |
Specifications
191-24-2 | |
100.0 | |
Yellow | |
Authentic | |
Glass bottle | |
MFCD00004135 | |
benzo ghi perylene, 1,12-benzoperylene, 1,12-benzperylene, benzo g,h,i perylene, benzo ghi pyrilene, benzo-1,12-perylene, unii-5zme2e2q9l, ccris 784 | |
GYFAGKUZYNFMBN-UHFFFAOYSA-N | |
276.33 | |
CHEBI:34568 | |
98+% | |
Benzo[ghi]perylene, 98+% |
98.0 | |
276.0°C to 280.0°C | |
>500.0°C | |
98+% | |
C22H12 | |
25 mg | |
Solubility in water: insoluble | |
C1=CC2=C3C(=C1)C4=CC=CC5=C4C6=C(C=C5)C=CC(=C36)C=C2 | |
9117 | |
276.33 | |
Needle-like Crystals or Crystalline Powder |
Safety and Handling
GHS H Statement
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid release to the environment.
GHS Signal Word: Warning
EINECSNumber : 205-883-8
RTECSNumber : DI6200500
RUO – Research Use Only