missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzothiazole-6-carboxylic acid, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 439700050

 View more versions of this product

Catalog No. AC439700050



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Benzothiazole-6-carboxylic acid
95.0
241°C to 243°C
Authentic
Glass Bottle
MFCD00111651
benzothiazole-6-carboxylic acid, 6-benzothiazolecarboxylic acid, benzo d thiazole-6-carboxylic acid, zlchem 73, pubchem9871, enamine_005417, benzothiazole-6-carboxylic, 6-benzothiazolecarboxylicacid, 6-benzothiazole carboxylic acid, cl-cob-iii-274-2
OC(=O)C1=CC=C2N=CSC2=C1
179.19
179.2
Powder
3622-35-3
100.0
Brown to White
95% min. (HPLC)
C8H5NO2S
5 g
DMPZJACLHDWUFS-UHFFFAOYSA-N
1,3-benzothiazole-6-carboxylic acid
601670
96%

Chemical Identifiers

3622-35-3
179.19
DMPZJACLHDWUFS-UHFFFAOYSA-N
601670
OC(=O)C1=CC=C2N=CSC2=C1
C8H5NO2S
MFCD00111651
benzothiazole-6-carboxylic acid, 6-benzothiazolecarboxylic acid, benzo d thiazole-6-carboxylic acid, zlchem 73, pubchem9871, enamine_005417, benzothiazole-6-carboxylic, 6-benzothiazolecarboxylicacid, 6-benzothiazole carboxylic acid, cl-cob-iii-274-2
1,3-benzothiazole-6-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only