Learn More
Benzothiazole-6-carboxylic acid, 96%, Thermo Scientific Chemicals
$227.76 - $227.76
Chemical Identifiers
CAS | 3622-35-3 |
---|---|
Molecular Formula | C8H5NO2S |
Molecular Weight (g/mol) | 179.19 |
MDL Number | MFCD00111651 |
InChI Key | DMPZJACLHDWUFS-UHFFFAOYSA-N |
Synonym | benzothiazole-6-carboxylic acid, 6-benzothiazolecarboxylic acid, benzo d thiazole-6-carboxylic acid, zlchem 73, pubchem9871, enamine_005417, benzothiazole-6-carboxylic, 6-benzothiazolecarboxylicacid, 6-benzothiazole carboxylic acid, cl-cob-iii-274-2 |
PubChem CID | 601670 |
IUPAC Name | 1,3-benzothiazole-6-carboxylic acid |
SMILES | OC(=O)C1=CC=C2N=CSC2=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC439700050
|
Thermo Scientific Chemicals
439700050 |
5 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3622-35-3 | |
179.19 | |
DMPZJACLHDWUFS-UHFFFAOYSA-N | |
601670 | |
OC(=O)C1=CC=C2N=CSC2=C1 |
C8H5NO2S | |
MFCD00111651 | |
benzothiazole-6-carboxylic acid, 6-benzothiazolecarboxylic acid, benzo d thiazole-6-carboxylic acid, zlchem 73, pubchem9871, enamine_005417, benzothiazole-6-carboxylic, 6-benzothiazolecarboxylicacid, 6-benzothiazole carboxylic acid, cl-cob-iii-274-2 | |
1,3-benzothiazole-6-carboxylic acid |
Specifications
3622-35-3 | |
100.0 | |
Brown to White | |
95% min. (HPLC) | |
C8H5NO2S | |
5 g | |
DMPZJACLHDWUFS-UHFFFAOYSA-N | |
1,3-benzothiazole-6-carboxylic acid | |
601670 | |
96% | |
Benzothiazole-6-carboxylic acid |
95.0 | |
241°C to 243°C | |
Authentic | |
Glass Bottle | |
MFCD00111651 | |
benzothiazole-6-carboxylic acid, 6-benzothiazolecarboxylic acid, benzo d thiazole-6-carboxylic acid, zlchem 73, pubchem9871, enamine_005417, benzothiazole-6-carboxylic, 6-benzothiazolecarboxylicacid, 6-benzothiazole carboxylic acid, cl-cob-iii-274-2 | |
OC(=O)C1=CC=C2N=CSC2=C1 | |
179.19 | |
179.2 | |
Powder |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
Warning
RUO – Research Use Only