missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl 3-pyrroline-1-carboxylate, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 371690050

 View more versions of this product

Catalog No. AC371690050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Benzyl 3-pyrroline-1-carboxylate
97.5
1.1320g/mL
>110°C
97.5% min. (GC)
C12H13NO2
5g
benzyl 2,5-dihydro-1h-pyrrole-1-carboxylate, benzyl 3-pyrroline-1-carboxylate, 1-cbz-2,5-dihydro-1h-pyrrole, n-cbz-2,5-dihydropyrrole, n-cbz-3-pyrroline, n-carbobenzoxy-3-pyrroline, 2,5-dihydropyrrole-1-carboxylic acid benzyl ester, benzyl 2h-pyrrole-1 5h-carboxylate, n-benzyloxycarbonyl-3-pyrroline, 1h-pyrrole-1-carboxylic acid, 2,5-dihydro-, phenylmethyl ester
C1C=CCN1C(=O)OCC2=CC=CC=C2
203.24
203.24
31970-04-4
100.0
133.0°C to 137.0°C (0.6mmHg)
Authentic
Glass bottle
1.544
1.132
XSKKIFJNZPNVGO-UHFFFAOYSA-N
benzyl 2,5-dihydropyrrole-1-carboxylate
643471
98%

Chemical Identifiers

31970-04-4
203.24
benzyl 2,5-dihydro-1h-pyrrole-1-carboxylate, benzyl 3-pyrroline-1-carboxylate, 1-cbz-2,5-dihydro-1h-pyrrole, n-cbz-2,5-dihydropyrrole, n-cbz-3-pyrroline, n-carbobenzoxy-3-pyrroline, 2,5-dihydropyrrole-1-carboxylic acid benzyl ester, benzyl 2h-pyrrole-1 5h-carboxylate, n-benzyloxycarbonyl-3-pyrroline, 1h-pyrrole-1-carboxylic acid, 2,5-dihydro-, phenylmethyl ester
benzyl 2,5-dihydropyrrole-1-carboxylate
C12H13NO2
XSKKIFJNZPNVGO-UHFFFAOYSA-N
643471
C1C=CCN1C(=O)OCC2=CC=CC=C2
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and k

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only