missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl 3-pyrroline-1-carboxylate, 98%, Thermo Scientific™

Chemical Identifiers

CAS 31970-04-4
Molecular Formula C12H13NO2
Molecular Weight (g/mol) 203.24
InChI Key XSKKIFJNZPNVGO-UHFFFAOYSA-N
Synonym benzyl 2,5-dihydro-1h-pyrrole-1-carboxylate, benzyl 3-pyrroline-1-carboxylate, 1-cbz-2,5-dihydro-1h-pyrrole, n-cbz-2,5-dihydropyrrole, n-cbz-3-pyrroline, n-carbobenzoxy-3-pyrroline, 2,5-dihydropyrrole-1-carboxylic acid benzyl ester, benzyl 2h-pyrrole-1 5h-carboxylate, n-benzyloxycarbonyl-3-pyrroline, 1h-pyrrole-1-carboxylic acid, 2,5-dihydro-, phenylmethyl ester
PubChem CID 643471
IUPAC Name benzyl 2,5-dihydropyrrole-1-carboxylate
SMILES C1C=CCN1C(=O)OCC2=CC=CC=C2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC371690050
SDS View Documents
Thermo Scientific Chemicals
371690050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

31970-04-4
203.24
benzyl 2,5-dihydro-1h-pyrrole-1-carboxylate, benzyl 3-pyrroline-1-carboxylate, 1-cbz-2,5-dihydro-1h-pyrrole, n-cbz-2,5-dihydropyrrole, n-cbz-3-pyrroline, n-carbobenzoxy-3-pyrroline, 2,5-dihydropyrrole-1-carboxylic acid benzyl ester, benzyl 2h-pyrrole-1 5h-carboxylate, n-benzyloxycarbonyl-3-pyrroline, 1h-pyrrole-1-carboxylic acid, 2,5-dihydro-, phenylmethyl ester
benzyl 2,5-dihydropyrrole-1-carboxylate
C12H13NO2
XSKKIFJNZPNVGO-UHFFFAOYSA-N
643471
C1C=CCN1C(=O)OCC2=CC=CC=C2

Specifications

31970-04-4
100.0
133.0°C to 137.0°C (0.6mmHg)
Authentic
Glass bottle
1.544
1.132
XSKKIFJNZPNVGO-UHFFFAOYSA-N
benzyl 2,5-dihydropyrrole-1-carboxylate
643471
98%
97.5
1.1320g/mL
>110°C
97.5% min. (GC)
C12H13NO2
5g
benzyl 2,5-dihydro-1h-pyrrole-1-carboxylate, benzyl 3-pyrroline-1-carboxylate, 1-cbz-2,5-dihydro-1h-pyrrole, n-cbz-2,5-dihydropyrrole, n-cbz-3-pyrroline, n-carbobenzoxy-3-pyrroline, 2,5-dihydropyrrole-1-carboxylic acid benzyl ester, benzyl 2h-pyrrole-1 5h-carboxylate, n-benzyloxycarbonyl-3-pyrroline, 1h-pyrrole-1-carboxylic acid, 2,5-dihydro-, phenylmethyl ester
C1C=CCN1C(=O)OCC2=CC=CC=C2
203.24
203.24
Benzyl 3-pyrroline-1-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and k

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only