missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl 4-bromopiperidine-1-carboxylate, 96%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 427030010

 View more versions of this product

Catalog No. AC427030010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Benzyl 4-bromopiperidine-1-carboxylate
95.0
1.3740g/mL
>110°C
Glass bottle
MFCD02681990
1.374
BFWKSPOSMPKTSV-UHFFFAOYSA-N
benzyl 4-bromopiperidine-1-carboxylate
2776275
96%
166953-64-6
100.0
144°C (0.3mmHg)
95% min. (HPLC)
C13H16BrNO2
1g
benzyl 4-bromotetrahydro-1 2h-pyridinecarboxylate, n-cbz-4-bromo-piperidine, 4-bromo-n-z-piperidine, 4-bromopiperidine, n-cbz protected, 1-cbz-4-bromopiperidine, benzyl4-bromopiperidine-1-carboxylate, benzyl 4-bromo-1-piperidinecarboxylate, pubchem16157, buttpark 9350-52, 4-bromo-n-cbz piperidine
C1CN(CCC1Br)C(=O)OCC2=CC=CC=C2
298.18
298.18

Chemical Identifiers

166953-64-6
298.18
BFWKSPOSMPKTSV-UHFFFAOYSA-N
2776275
C1CN(CCC1Br)C(=O)OCC2=CC=CC=C2
C13H16BrNO2
MFCD02681990
benzyl 4-bromotetrahydro-1 2h-pyridinecarboxylate, n-cbz-4-bromo-piperidine, 4-bromo-n-z-piperidine, 4-bromopiperidine, n-cbz protected, 1-cbz-4-bromopiperidine, benzyl4-bromopiperidine-1-carboxylate, benzyl 4-bromo-1-piperidinecarboxylate, pubchem16157, buttpark 9350-52, 4-bromo-n-cbz piperidine
benzyl 4-bromopiperidine-1-carboxylate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only