missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl 4-bromopiperidine-1-carboxylate, 96%, Thermo Scientific™

Chemical Identifiers

CAS 166953-64-6
Molecular Formula C13H16BrNO2
Molecular Weight (g/mol) 298.18
MDL Number MFCD02681990
InChI Key BFWKSPOSMPKTSV-UHFFFAOYSA-N
Synonym benzyl 4-bromotetrahydro-1 2h-pyridinecarboxylate, n-cbz-4-bromo-piperidine, 4-bromo-n-z-piperidine, 4-bromopiperidine, n-cbz protected, 1-cbz-4-bromopiperidine, benzyl4-bromopiperidine-1-carboxylate, benzyl 4-bromo-1-piperidinecarboxylate, pubchem16157, buttpark 9350-52, 4-bromo-n-cbz piperidine
PubChem CID 2776275
IUPAC Name benzyl 4-bromopiperidine-1-carboxylate
SMILES C1CN(CCC1Br)C(=O)OCC2=CC=CC=C2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC427030010
SDS View Documents
Thermo Scientific Chemicals
427030010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

166953-64-6
298.18
BFWKSPOSMPKTSV-UHFFFAOYSA-N
2776275
C1CN(CCC1Br)C(=O)OCC2=CC=CC=C2
C13H16BrNO2
MFCD02681990
benzyl 4-bromotetrahydro-1 2h-pyridinecarboxylate, n-cbz-4-bromo-piperidine, 4-bromo-n-z-piperidine, 4-bromopiperidine, n-cbz protected, 1-cbz-4-bromopiperidine, benzyl4-bromopiperidine-1-carboxylate, benzyl 4-bromo-1-piperidinecarboxylate, pubchem16157, buttpark 9350-52, 4-bromo-n-cbz piperidine
benzyl 4-bromopiperidine-1-carboxylate

Specifications

166953-64-6
100.0
144°C (0.3mmHg)
95% min. (HPLC)
C13H16BrNO2
1g
benzyl 4-bromotetrahydro-1 2h-pyridinecarboxylate, n-cbz-4-bromo-piperidine, 4-bromo-n-z-piperidine, 4-bromopiperidine, n-cbz protected, 1-cbz-4-bromopiperidine, benzyl4-bromopiperidine-1-carboxylate, benzyl 4-bromo-1-piperidinecarboxylate, pubchem16157, buttpark 9350-52, 4-bromo-n-cbz piperidine
C1CN(CCC1Br)C(=O)OCC2=CC=CC=C2
298.18
298.18
Benzyl 4-bromopiperidine-1-carboxylate
95.0
1.3740g/mL
>110°C
Glass bottle
MFCD02681990
1.374
BFWKSPOSMPKTSV-UHFFFAOYSA-N
benzyl 4-bromopiperidine-1-carboxylate
2776275
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only