missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl acetoacetate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1491522

 View more versions of this product

Catalog No. AAA1491522


Explore available promotions

Description

Description

Benzyl acetoacetate is used as a Pharmaceutical and fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl acetoacetate is used as a Pharmaceutical and fine chemical intermediate.

Solubility
It is soluble in alkali solutions.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications

Specifications

Benzyl acetoacetate
1.113
108°C (226°F)
1.512
100 g
benzyl acetoacetate, acetoacetic acid benzyl ester, benzyl acetylacetate, acetoacetic acid, benzyl ester, phenylmethyl 3-oxobutanoate, butanoic acid, 3-oxo-, phenylmethyl ester, benzyl beta-ketobutyrate, ac-benzyl, unii-f9s0xgv18x, f9s0xgv18x
WOFAGNLBCJWEOE-UHFFFAOYSA-N
benzyl 3-oxobutanoate
142266
97%
5396-89-4
276°C
C11H12O3
MFCD00008785
2049615
It is soluble in alkali solutions.
CC(=O)CC(=O)OCC1=CC=CC=C1
192.214
192.22

Chemical Identifiers

5396-89-4
192.214
WOFAGNLBCJWEOE-UHFFFAOYSA-N
142266
CC(=O)CC(=O)OCC1=CC=CC=C1
C11H12O3
MFCD00008785
benzyl acetoacetate, acetoacetic acid benzyl ester, benzyl acetylacetate, acetoacetic acid, benzyl ester, phenylmethyl 3-oxobutanoate, butanoic acid, 3-oxo-, phenylmethyl ester, benzyl beta-ketobutyrate, ac-benzyl, unii-f9s0xgv18x, f9s0xgv18x
benzyl 3-oxobutanoate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280i-P305+P351+P338

H319

EINECSNumber : 226-416-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only