missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Benzyl acetoacetate, 97%, Thermo Scientific Chemicals
$49.00 - $135.23
Chemical Identifiers
CAS | 5396-89-4 |
---|---|
Molecular Formula | C11H12O3 |
Molecular Weight (g/mol) | 192.214 |
MDL Number | MFCD00008785 |
InChI Key | WOFAGNLBCJWEOE-UHFFFAOYSA-N |
Synonym | benzyl acetoacetate, acetoacetic acid benzyl ester, benzyl acetylacetate, acetoacetic acid, benzyl ester, phenylmethyl 3-oxobutanoate, butanoic acid, 3-oxo-, phenylmethyl ester, benzyl beta-ketobutyrate, ac-benzyl, unii-f9s0xgv18x, f9s0xgv18x |
PubChem CID | 142266 |
IUPAC Name | benzyl 3-oxobutanoate |
SMILES | CC(=O)CC(=O)OCC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1491514
|
Thermo Scientific Chemicals
A1491514 |
25 g |
Each for $49.00
|
|
|||||
AAA1491518
|
Thermo Scientific Chemicals
A1491518 |
50 g |
Each for $83.11
|
|
|||||
AAA1491522
|
Thermo Scientific Chemicals
A1491522 |
100 g |
Each for $135.23
|
|
|||||
Description
Benzyl acetoacetate is used as a Pharmaceutical and fine chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzyl acetoacetate is used as a Pharmaceutical and fine chemical intermediate.
Solubility
It is soluble in alkali solutions.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Chemical Identifiers
5396-89-4 | |
192.214 | |
WOFAGNLBCJWEOE-UHFFFAOYSA-N | |
142266 | |
CC(=O)CC(=O)OCC1=CC=CC=C1 |
C11H12O3 | |
MFCD00008785 | |
benzyl acetoacetate, acetoacetic acid benzyl ester, benzyl acetylacetate, acetoacetic acid, benzyl ester, phenylmethyl 3-oxobutanoate, butanoic acid, 3-oxo-, phenylmethyl ester, benzyl beta-ketobutyrate, ac-benzyl, unii-f9s0xgv18x, f9s0xgv18x | |
benzyl 3-oxobutanoate |
Specifications
5396-89-4 | |
276°C | |
C11H12O3 | |
MFCD00008785 | |
2049615 | |
It is soluble in alkali solutions. | |
CC(=O)CC(=O)OCC1=CC=CC=C1 | |
192.214 | |
192.22 | |
Benzyl acetoacetate |
1.113 | |
108°C (226°F) | |
1.512 | |
25 g | |
benzyl acetoacetate, acetoacetic acid benzyl ester, benzyl acetylacetate, acetoacetic acid, benzyl ester, phenylmethyl 3-oxobutanoate, butanoic acid, 3-oxo-, phenylmethyl ester, benzyl beta-ketobutyrate, ac-benzyl, unii-f9s0xgv18x, f9s0xgv18x | |
WOFAGNLBCJWEOE-UHFFFAOYSA-N | |
benzyl 3-oxobutanoate | |
142266 | |
97% |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280i-P305+P351+P338
H319
EINECSNumber : 226-416-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only